Thermo Scientific™

Methyl sulfoxide-d6, for NMR, 99.9% atom D, Thermo Scientific Chemicals

Catalog number: 166290101
10 mL, Septum Bottle, Each
Thermo Scientific™

Methyl sulfoxide-d6, for NMR, 99.9% atom D, Thermo Scientific Chemicals

Catalog number: 166290101
10 mL, Septum Bottle, Each
Quantity
Packaging
Catalog number: 166290101
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2206-27-1
IUPAC Name
(²H₃)methanesulfinyl(²H₃)methane
Molecular Formula
C2H6OS
InChI Key
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Appearance (Color)
Clear colorless
D-Enrichment
>=99.85 %
Water
=<0.03 % (K.F.)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text