Thermo Scientific Chemicals

Isocarbostyril, 98%, Thermo Scientific Chemicals

Catalog number: 166460050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Isocarbostyril, 98%, Thermo Scientific Chemicals

Catalog number: 166460050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 166460050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
491-30-5
IUPAC Name
1,2-dihydroisoquinolin-1-one
Molecular Formula
C9H7NO
InChI Key
VDBNYAPERZTOOF-UHFFFAOYSA-N
SMILES
O=C1NC=CC2=CC=CC=C12
HPLC
>=97.5 %
Appearance (Color)
White to light brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
210°C to 215°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text