Thermo Scientific Chemicals

Taurine, 99%, Thermo Scientific Chemicals

Catalog number: 166541000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Taurine, 99%, Thermo Scientific Chemicals

Catalog number: 166541000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 166541000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
107-35-7
IUPAC Name
2-aminoethane-1-sulfonic acid
Molecular Formula
C2H7NO3S
InChI Key
XOAAWQZATWQOTB-UHFFFAOYSA-N
SMILES
NCCS(O)(=O)=O
Titration with NaOH
>=98.5 %
Chloride (Cl)
=<110 ppm
Sulfate (SO4)
=<140 ppm
Ammonium (NH4)
=<200 ppm
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text