Taurine, 99%
Taurine, 99%
Taurine, 99%
Thermo Scientific Chemicals

Taurine, 99%

CAS: 107-35-7 | C2H7NO3S | 125.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 166545000
Price (EUR)
99,45
Online Exclusive
117,00
Save 17,55 (15%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
99,45
Online Exclusive
117,00
Save 17,55 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Name2-aminoethane-1-sulfonic acid
Molecular FormulaC2H7NO3S
InChI KeyXOAAWQZATWQOTB-UHFFFAOYSA-N
SMILESNCCS(O)(=O)=O
Molecular Weight (g/mol)125.14
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Appearance (Color)White
Arsenic (As)=<2 ppm
Titration with NaOH>=98.5 %
Chloride (Cl)=<110 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link