2-Methyl-2-butanol, 99%, pure
2-Methyl-2-butanol, 99%, pure
2-Methyl-2-butanol, 99%, pure
2-Methyl-2-butanol, 99%, pure
Thermo Scientific Chemicals

2-Methyl-2-butanol, 99%, pure

CAS: 75-85-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Catalog number 166620010
Price (USD)
117.65
Special offer
Online exclusive
138.00
Save 20.35 (15%)
Each
In stock
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)
117.65
Special offer
Online exclusive
138.00
Save 20.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS75-85-4
IUPAC Name2-methylbutan-2-ol
Molecular FormulaC5H12O
InChI KeyMSXVEPNJUHWQHW-UHFFFAOYSA-N
SMILESCCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.1 %
Refractive index1.4040 to 1.4060 (20°C, 589 nm)
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link