2-Methyl-2-butanol, 99%, pure, Thermo Scientific Chemicals
2-Methyl-2-butanol, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-2-butanol, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Catalog number 166620100
Price (USD)/ Each
408.65
Online exclusive
454.00 
Save 45.35 (10%)
-
Add to cart
Quantity:
10 L
Request bulk or custom format
Price (USD)/ Each
408.65
Online exclusive
454.00 
Save 45.35 (10%)
Add to cart
2-Methyl-2-butanol, 99%, pure, Thermo Scientific Chemicals
Catalog number166620100
Price (USD)/ Each
408.65
Online exclusive
454.00 
Save 45.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-85-4
IUPAC Name2-methylbutan-2-ol
Molecular FormulaC5H12O
InChI KeyMSXVEPNJUHWQHW-UHFFFAOYSA-N
SMILESCCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Water=<0.1 %
Refractive index1.4040 to 1.4060 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only