5-Bromo-2-thiophenecarboxaldehyde, 97%, Thermo Scientific Chemicals
5-Bromo-2-thiophenecarboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-thiophenecarboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
25 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 166651000
Price (USD)/ Each
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name5-bromothiophene-2-carbaldehyde
Molecular FormulaC5H3BrOS
InChI KeyGFBVUFQNHLUCPX-UHFFFAOYSA-N
SMILESBrC1=CC=C(S1)C=O
Molecular Weight (g/mol)191.04
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.6380 to 1.6410 (20°C, 589 nm)
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
Appearance (Form)Liquid after melting
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only