Thermo Scientific Chemicals

N-Acetyl-4-piperidone, 97%, Thermo Scientific Chemicals

Catalog number: 166821000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Acetyl-4-piperidone, 97%, Thermo Scientific Chemicals

Catalog number: 166821000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 166821000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32161-06-1
IUPAC Name
1-acetylpiperidin-4-one
Molecular Formula
C7H11NO2
InChI Key
NNFOVLFUGLWWCL-UHFFFAOYSA-N
SMILES
CC(=O)N1CCC(=O)CC1
Appearance (Color)
Clear orange
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.5015 to 1.5035 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text