3-Amino-4-pyrazolecarbonitrile, 98%, Thermo Scientific Chemicals
3-Amino-4-pyrazolecarbonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-4-pyrazolecarbonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Packaging:
Glass bottle
Catalog number 166850250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16617-46-2
IUPAC Name5-amino-1H-pyrazole-4-carbonitrile
Molecular FormulaC4H4N4
InChI KeyFFNKBQRKZRMYCL-UHFFFAOYSA-N
SMILESNC1=C(C=NN1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to orange-brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point172°C to 176°C
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only