Thermo Scientific Chemicals

4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 166965000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 166965000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 166965000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-88-3
IUPAC Name
2-(4-chlorophenoxy)acetic acid
Molecular Formula
C8H7ClO3
InChI Key
SODPIMGUZLOIPE-UHFFFAOYSA-N
SMILES
OC(=O)COC1=CC=C(Cl)C=C1
Appearance (Color)
White to light beige
Melting point
>=156°C
Water
=<1 %
Titration with NaOH
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text