Thermo Scientific Chemicals

N-(Trimethylsilyl)imidazole, 97%, Thermo Scientific Chemicals

Catalog number: 167131000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(Trimethylsilyl)imidazole, 97%, Thermo Scientific Chemicals

Catalog number: 167131000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 167131000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18156-74-6
IUPAC Name
1-(trimethylsilyl)-1H-imidazole
Molecular Formula
C6H12N2Si
InChI Key
YKFRUJSEPGHZFJ-UHFFFAOYSA-N
SMILES
C[Si](C)(C)N1C=CN=C1
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=96.0 %
Refractive index
1.4750 to 1.4780 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text