Thermo Scientific Chemicals

Diphenylcarbamyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 167141000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

Diphenylcarbamyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 167141000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 167141000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
83-01-2
IUPAC Name
N,N-diphenylcarbamoyl chloride
Molecular Formula
C13H10ClNO
InChI Key
XNBKKRFABABBPM-UHFFFAOYSA-N
SMILES
ClC(=O)N(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Crystalline powder
Appearance (Color)
Beige to grey-green
Infrared spectrum
Conforms
Melting point
81°C to 85°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text