Thermo Scientific Chemicals

N-Carbethoxy-4-piperidone, 98.5+%, Thermo Scientific Chemicals

Catalog number: 167240100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Carbethoxy-4-piperidone, 98.5+%, Thermo Scientific Chemicals

Catalog number: 167240100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 167240100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
29976-53-2
IUPAC Name
ethyl 4-oxopiperidine-1-carboxylate
Molecular Formula
C8H13NO3
InChI Key
LUBGFMZTGFXIIN-UHFFFAOYSA-N
SMILES
CCOC(=O)N1CCC(=O)CC1
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4740 to 1.4760 (20°C, 589 nm)
Appearance
May darken on storage
Water
=<0.5 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text