Decanoic acid, 99%
Decanoic acid, 99%
Decanoic acid, 99%
Thermo Scientific Chemicals

Decanoic acid, 99%

CAS: 334-48-5 | C10H20O2 | 172.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 167270010
Price (USD)
130.65
Special offer
Online exclusive
Ends: 26-Sep-2025
162.00
Save 31.35 (19%)
Each
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
Ends: 26-Sep-2025
162.00
Save 31.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS334-48-5
IUPAC Namedecanoic acid
Molecular FormulaC10H20O2
InChI KeyGHVNFZFCNZKVNT-UHFFFAOYSA-N
SMILESCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals and/or chunks or fused solid or liquid
Melting point29°C to 34°C
Infrared spectrumConforms
Appearance (Color)Colorless or white to cream
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link