Thermo Scientific Chemicals

Lauric acid, 99%

Catalog number: 167280050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Lauric acid, 99%

Catalog number: 167280050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 167280050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-07-7
IUPAC Name
dodecanoic acid
Molecular Formula
C12H24O2
InChI Key
POULHZVOKOAJMA-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCC(O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder or flakes
GC
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text