Thermo Scientific Chemicals

2-Adamantanamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 167300050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Adamantanamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 167300050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 167300050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10523-68-9
IUPAC Name
hydrogen adamantan-2-amine chloride
Molecular Formula
C10H18ClN
InChI Key
WLDWDRZITJEWRJ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC1C2CC3CC(C2)CC1C3
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals
Appearance (Color)
White
Titration with HClO4
98.0 to 102.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text