2-Adamantanamine hydrochloride, 99%, Thermo Scientific Chemicals
2-Adamantanamine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Adamantanamine hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 167300250
Price (USD)/ Each
423.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
423.00
Add to cart
2-Adamantanamine hydrochloride, 99%, Thermo Scientific Chemicals
Catalog number167300250
Price (USD)/ Each
423.00
-
Add to cart
Chemical Identifiers
CAS10523-68-9
IUPAC Namehydrogen adamantan-2-amine chloride
Molecular FormulaC10H18ClN
InChI KeyWLDWDRZITJEWRJ-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1C2CC3CC(C2)CC1C3
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder or crystals
Appearance (Color)White
Titration with HClO498.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only