2,6-Di-tert-butyl-p-benzoquinone, 98%, Thermo Scientific Chemicals
2,6-Di-tert-butyl-p-benzoquinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Di-tert-butyl-p-benzoquinone, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 167340010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS719-22-2
IUPAC Name2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione
Molecular FormulaC14H20O2
InChI KeyRDQSIADLBQFVMY-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Yellow to orange
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals
Melting point64°C to 70°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only