Thermo Scientific Chemicals

2,6-Di-tert-butyl-p-benzoquinone, 98%, Thermo Scientific Chemicals

Catalog number: 167340050
5 g, Each
Thermo Scientific Chemicals

2,6-Di-tert-butyl-p-benzoquinone, 98%, Thermo Scientific Chemicals

Catalog number: 167340050
5 g, Each
Quantity
Catalog number: 167340050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
719-22-2
IUPAC Name
2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione
Molecular Formula
C14H20O2
InChI Key
RDQSIADLBQFVMY-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C
Infrared spectrum
Conforms
Appearance (Color)
Yellow to orange
GC
>=97.5 %
Appearance (Form)
Crystalline powder or crystals
Melting point
64°C to 70°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text