Thermo Scientific Chemicals

3-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 167360250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 167360250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 167360250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2312-23-4
IUPAC Name
hydrogen (3-chlorophenyl)hydrazine chloride
Molecular Formula
C6H8Cl2N2
InChI Key
CRRIAWUJYMLJOE-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=CC(Cl)=C1
Appearance (Color)
White to light brown to pink
Titration Argentometric
96.0 to 104.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text