Thermo Scientific Chemicals

4-Bromo-2-chloroaniline, 99%, Thermo Scientific Chemicals

Catalog number: 167480100
10 g, Each
Thermo Scientific Chemicals

4-Bromo-2-chloroaniline, 99%, Thermo Scientific Chemicals

Catalog number: 167480100
10 g, Each
Quantity
Catalog number: 167480100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38762-41-3
IUPAC Name
3-bromo-5-chloroaniline
Molecular Formula
C6H5BrClN
InChI Key
YYBRLVWWPRAQDX-UHFFFAOYSA-N
SMILES
NC1=CC(Cl)=CC(Br)=C1
Appearance (Form)
Crystalline powder
Appearance (Color)
Light yellow to light brown
Melting point
67°C to 70°C
Infrared spectrum
Conforms
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text