2,6-Pyridinedimethanol, 98%, Thermo Scientific Chemicals
2,6-Pyridinedimethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Pyridinedimethanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 167551000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name[6-(hydroxymethyl)pyridin-2-yl]methanol
Molecular FormulaC7H9NO2
InChI KeyWWFMINHWJYHXHF-UHFFFAOYSA-N
SMILESOCC1=CC=CC(CO)=N1
Molecular Weight (g/mol)139.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to yellow to beige
GC>=97.5 %
Infrared spectrumConforms
Melting point108°C to 115°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only