96 Well Edge Plate, TC Surface, No Lid, Pack of 10
Tired of paying too much for ECM plates? Try these ECM plates that are easy on your budget »
96 Well Edge Plate, TC Surface, No Lid, Pack of 10
Thermo Scientific™
96 Well Edge Plate, TC Surface, No Lid, Pack of 10
Green features
Color:Clear; Sterility:Sterile; Lid:Without Lid
Have Questions?
Catalog number 167554
Price (USD)/ Case of 160
360.00
-
Add to cart
Price (USD)/ Case of 160
360.00
Add to cart
96 Well Edge Plate, TC Surface, No Lid, Pack of 10
Catalog number167554
Price (USD)/ Case of 160
360.00
-
Add to cart
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Features
  • The Edge 2.0 Plate provides an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect.
  • The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL sterile water, buffer or media.
  • Replenish fluid in the reservoirs every 3 days in the humidified incubator to ensure the effectiveness of the evaporation barrier.
  • Nunclon™ Delta cell culture surface treatment of the wells promotes cell attachment and growth.
  • The non-treated surface is available for suspension cell culture or general solution-based assays
  • Use the Edge 2.0 Plate in conjunction with the Thermo Scientific™ Varioskan™ LUX multimode microplate reader. The filled moat of the plate functions as passive humidity control, allowing for simultaneous incubation and signal detection during long-term applications with living cells inside the plate reader.
Warranty: 90 days
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
CertifiedNunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
LidWithout Lid
Culture Area0.35 cm2/well
Array8 x 12
Packaging10 Pk., 160 Cases
Product LineNunc
ColorClear
Product TypeMicroplate
Skirt StyleLow Flange
MaterialPolystyrene
Volume (Metric) Well400 μL
No. of Wells96
SterilitySterile
Surface TreatmentNunclon Delta
Well ShapeFlat Bottom (F-well)
Green FeaturesLess waste, fewer resources used
No. per Case160
No. per Pack10
Quantity160 Plates
Unit SizeCase of 160