Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Thermo Scientific™
Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate.
Have Questions?
Catalog NumberQuantity
167574Case of 50
Catalog number 167574
Price (USD)
139.65
Online Exclusive
149.00
Save 9.35 (6%)
Case of 50
Quantity:
Case of 50
Price (USD)
139.65
Online Exclusive
149.00
Save 9.35 (6%)
Case of 50
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
CertifiedNunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
Culture Area0.35 cm2/well
LidWithout Lid
No. per Case50
No. per Pack1
Packaging1/Pk., 50/Cs.
Product TypeMicroplate
QuantityCase of 50
Skirt StyleLow Flange
Array8 x 12
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNunclon Delta
Volume (Metric) Well400 μL
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 50