Thermo Scientific Chemicals

Benzonitrile, 99+%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 167670010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Benzonitrile, 99+%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 167670010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 167670010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
100-47-0
IUPAC Name
benzonitrile
Molecular Formula
C7H5N
InChI Key
JFDZBHWFFUWGJE-UHFFFAOYSA-N
SMILES
N#CC1=CC=CC=C1
Water
=<0.03 %
Appearance (Form)
Clear liquid
UV
at 300 nm A: =<1
UV
at 400 nm A: =<0.01
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text