Benzonitrile, 99+%, for spectroscopy, Thermo Scientific Chemicals
Benzonitrile, 99+%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzonitrile, 99+%, for spectroscopy, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
1 L
Packaging:
Glass bottle
Catalog number 167672500
Price (USD)/ Each
82.65
Online exclusive
91.60 
Save 8.95 (10%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
82.65
Online exclusive
91.60 
Save 8.95 (10%)
Add to cart
Benzonitrile, 99+%, for spectroscopy, Thermo Scientific Chemicals
Catalog number167672500
Price (USD)/ Each
82.65
Online exclusive
91.60 
Save 8.95 (10%)
-
Add to cart
Chemical Identifiers
CAS100-47-0
IUPAC Namebenzonitrile
Molecular FormulaC7H5N
InChI KeyJFDZBHWFFUWGJE-UHFFFAOYSA-N
SMILESN#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.03 %
Appearance (Form)Clear liquid
UVat 300 nm A: =<1
UVat 400 nm A: =<0.01
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only