Thermo Scientific Chemicals

n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 167680010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 167680010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 167680010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-86-4
IUPAC Name
butyl acetate
Molecular Formula
C6H12O2
InChI Key
DKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILES
CCCCOC(C)=O
Appearance (Form)
Clear liquid
Water
=<0.05 %
UV
at 254 nm A: =<1
UV
at 260 nm A: =<0.15
UV
at 300 nm A: =<0.02

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text