2-Methyl-1-propanol, 99+%, spectrophotometric grade, Thermo Scientific Chemicals
2-Methyl-1-propanol, 99+%, spectrophotometric grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-1-propanol, 99+%, spectrophotometric grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
Glass bottle
Catalog number 167705000
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78-83-1
IUPAC Name2-methylpropan-1-ol
Molecular FormulaC4H10O
InChI KeyZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILESCC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 330 nm A: =<0.01
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.0 %
Water=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only