Thermo Scientific Chemicals

Carbon disulfide, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 167710250
25 mL, Glass Bottle, Each
Thermo Scientific Chemicals

Carbon disulfide, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 167710250
25 mL, Glass Bottle, Each
Quantity
Packaging
Catalog number: 167710250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-15-0
IUPAC Name
methanedithione
Molecular Formula
CS2
InChI Key
QGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILES
S=C=S
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
GC
>=99.85 %
Water
=<0.03 %
UV
at 380 nm A: =<1.3

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text