Dichloromethane, 99.8%, for spectroscopy, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.8%, for spectroscopy, stabilized with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethane, 99.8%, for spectroscopy, stabilized with amylene, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number 167775000
Price (USD)/ Each
51.65
Online exclusive
57.50 
Save 5.85 (10%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
51.65
Online exclusive
57.50 
Save 5.85 (10%)
Add to cart
Dichloromethane, 99.8%, for spectroscopy, stabilized with amylene, Thermo Scientific Chemicals
Catalog number167775000
Price (USD)/ Each
51.65
Online exclusive
57.50 
Save 5.85 (10%)
-
Add to cart
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
UVat 250 nm A: =<0.05
UVat 260 nm A: =<0.02
UVat 270 nm A: =<0.01
UVat 280 nm A: =<0.01
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only