Thermo Scientific Chemicals

n-heptane, for HPLC, ∽ 95% n-heptane, Thermo Scientific Chemicals

Catalog number: 167820025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

n-heptane, for HPLC, ∽ 95% n-heptane, Thermo Scientific Chemicals

Catalog number: 167820025
2.5 L, Glass Bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
142-82-5
IUPAC Name
heptane
Molecular Formula
C7H16
InChI Key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES
CCCCCCC
Appearance (Form)
Clear liquid
Water
=<0.01 %
UV
at 210 nm A: =<0.301
UV
1 cm cell vs HPLC-grade water
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 2
    Skin Corrosion/Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Aspiration Toxicity
    Category 1
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H304 - May be fatal if swallowed and enters airways

    H315 - Causes skin irritation

    H336 - May cause drowsiness or dizziness

    H410 - Very toxic to aquatic life with long lasting effects

    Physical Hazards

    H225 - Highly flammable liquid and vapor

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P273 - Avoid release to the environment

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P331 - Do NOT induce vomiting

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking