Methanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Methanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number 167835000
Price (USD)/ Each
26.65
Online exclusive
29.30 
Save 2.65 (9%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
26.65
Online exclusive
29.30 
Save 2.65 (9%)
Add to cart
Methanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Catalog number167835000
Price (USD)/ Each
26.65
Online exclusive
29.30 
Save 2.65 (9%)
-
Add to cart
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.05 % (Coulometric)
Residue after evaporation=<0.0003 %
Appearance (Form)Clear liquid
GC>=99.85 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only