Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Have Questions?
Catalog number 167850010
Price (USD)/ Each
150.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
177.00 
Save 26.35 (15%)
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
150.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
177.00 
Save 26.35 (15%)
Add to cart
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Catalog number167850010
Price (USD)/ Each
150.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
177.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
UVat 270 nm A: =<0.5
Water=<0.1 %
Appearance (Form)Clear liquid
GC>=99.85 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only