Isopropanol, 99+%, for spectroscopy, Thermo Scientific Chemicals
Isopropanol, 99+%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropanol, 99+%, for spectroscopy, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 167880010
Price (USD)/ Each
52.65
Online exclusive
58.10 
Save 5.45 (9%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
52.65
Online exclusive
58.10 
Save 5.45 (9%)
Add to cart
Isopropanol, 99+%, for spectroscopy, Thermo Scientific Chemicals
Catalog number167880010
Price (USD)/ Each
52.65
Online exclusive
58.10 
Save 5.45 (9%)
-
Add to cart
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
GC>=99.0 %
UVat 220 nm A: =<0.3
UVat 230 nm A: =<0.15
UVat 260 nm A: =<0.03
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only