Thermo Scientific Chemicals

Tetrachloroethylene, 99+%, spectrophotometric grade, Thermo Scientific Chemicals

Catalog number: 167890010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Tetrachloroethylene, 99+%, spectrophotometric grade, Thermo Scientific Chemicals

Catalog number: 167890010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 167890010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-18-4
IUPAC Name
tetrachloroethene
Molecular Formula
C2Cl4
InChI Key
CYTYCFOTNPOANT-UHFFFAOYSA-N
SMILES
ClC(Cl)=C(Cl)Cl
Water
=<0.05 %
UV
1 cm cell vs water
UV
at 305 nm A: =<0.1
UV
at 350 nm A: =<0.05
UV
at 400 nm A: =<0.03

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text