Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 167915000
500 mL, Glass Bottle, Each
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 167915000
500 mL, Glass Bottle, Each
Quantity
Packaging
Catalog number: 167915000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
540-84-1
IUPAC Name
2,2,4-trimethylpentane
Molecular Formula
C8H18
InChI Key
NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES
CC(C)CC(C)(C)C
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
GC
>=98.5 %
UV
at 230 nm A: =<0.1
UV
at 240 nm A: =<0.04

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text