Thermo Scientific Chemicals

Azidotrimethylsilane, 94%, Thermo Scientific Chemicals

Catalog number: 167930500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

Azidotrimethylsilane, 94%, Thermo Scientific Chemicals

Catalog number: 167930500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 167930500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4648-54-8
Appearance (Color)
Clear to slightly hazy colorless
Appearance (Form)
Liquid
GC
>=92 %
Refractive index
1.4110 to 1.4180 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transfer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text