Valeronitrile, 98%, Thermo Scientific Chemicals
Valeronitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Valeronitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number 167945000
Price (USD)/ Each
210.00
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
210.00
Add to cart
Valeronitrile, 98%, Thermo Scientific Chemicals
Catalog number167945000
Price (USD)/ Each
210.00
-
Add to cart
Chemical Identifiers
CAS110-59-8
IUPAC Namepentanenitrile
Molecular FormulaC5H9N
InChI KeyRFFFKMOABOFIDF-UHFFFAOYSA-N
SMILESCCCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.3960 to 1.3990 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only