Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%, Thermo Scientific Chemicals

Catalog number: 168005000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%, Thermo Scientific Chemicals

Catalog number: 168005000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 168005000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25561-30-2
IUPAC Name
trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
Molecular Formula
C8H18F3NOSi2
InChI Key
XCOBLONWWXQEBS-UHFFFAOYSA-N
SMILES
C[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=98.0 %
Refractive index
1.3829 to 1.3849 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text