2-Piperidinemethanol, 97%, Thermo Scientific Chemicals
2-Piperidinemethanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Piperidinemethanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 168030250
Price (USD)/ Each
174.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
174.00
Add to cart
2-Piperidinemethanol, 97%, Thermo Scientific Chemicals
Catalog number168030250
Price (USD)/ Each
174.00
-
Add to cart
Chemical Identifiers
CAS3433-37-2
IUPAC Name(2R)-2-(hydroxymethyl)piperidin-1-ium
Molecular FormulaC6H14NO
InChI KeyPRAYXGYYVXRDDW-ZCFIWIBFSA-O
SMILESOC[C@H]1CCCC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
Infrared spectrumConforms
Appearance (Color)White to light orange
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only