Thermo Scientific Chemicals

2,4,6-Tri(2-pyridyl)-s-triazine, 99%, Thermo Scientific Chemicals

Catalog number: 168071000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4,6-Tri(2-pyridyl)-s-triazine, 99%, Thermo Scientific Chemicals

Catalog number: 168071000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 168071000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3682-35-7
IUPAC Name
tris(pyridin-2-yl)-1,3,5-triazine
Molecular Formula
C18H12N6
InChI Key
KMVWNDHKTPHDMT-UHFFFAOYSA-N
SMILES
C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
Appearance (Color)
White to cream to yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
248°C to 252°C
Titration with HClO4
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text