trans-Styrylacetic acid, 96%, Thermo Scientific Chemicals
trans-Styrylacetic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-Styrylacetic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 168100010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(3E)-4-phenylbut-3-enoic acid
Molecular FormulaC10H10O2
InChI KeyPSCXFXNEYIHJST-QPJJXVBHSA-N
SMILESOC(=O)C\C=C\C1=CC=CC=C1
Molecular Weight (g/mol)162.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow-brown
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Melting point82°C to 88°C
HPLC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only