Thermo Scientific Chemicals

trans-Styrylacetic acid, 96%, Thermo Scientific Chemicals

Catalog number: 168100100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

trans-Styrylacetic acid, 96%, Thermo Scientific Chemicals

Catalog number: 168100100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 168100100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1914-58-5
IUPAC Name
(3E)-4-phenylbut-3-enoic acid
Molecular Formula
C10H10O2
InChI Key
PSCXFXNEYIHJST-QPJJXVBHSA-N
SMILES
OC(=O)C\C=C\C1=CC=CC=C1
Appearance (Color)
White to yellow-brown
Appearance (Form)
Crystalline powder or needles
Infrared spectrum
Conforms
Melting point
82°C to 88°C
HPLC
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text