Thermo Scientific Chemicals

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 168230010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 168230010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 168230010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17257-71-5
IUPAC Name
(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoate
Molecular Formula
C10H8F3O3
InChI Key
JJYKJUXBWFATTE-VIFPVBQESA-M
SMILES
CO[C@](C([O-])=O)(C1=CC=CC=C1)C(F)(F)F
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Viscous liquid or low melting solid
Infrared spectrum
Conforms
Refractive index
1.4720 to 1.4750 (20°C, 589 nm)
Titration with NaOH
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text