Thermo Scientific Chemicals

Methacrylic acid, 99.5%, extra pure, stabilized, Thermo Scientific Chemicals

Catalog number: 168310251
25 L, Metal Drum, Each
Thermo Scientific Chemicals

Methacrylic acid, 99.5%, extra pure, stabilized, Thermo Scientific Chemicals

Catalog number: 168310251
25 L, Metal Drum, Each
Quantity
Packaging
Catalog number: 168310251
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-41-4
IUPAC Name
2-methylprop-2-enoic acid
Molecular Formula
C4H6O2
InChI Key
CERQOIWHTDAKMF-UHFFFAOYSA-N
SMILES
CC(=C)C(O)=O
Appearance (Form)
Clear liquid
Refractive index
1.4295 to 1.4315 (20°C, 589 nm)
Stabilizer
ca. 250 ppm MEHQ
Water
=<0.2 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text