Methacrylic acid, 99.5%, extra pure, stabilized, Thermo Scientific Chemicals
Methacrylic acid, 99.5%, extra pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methacrylic acid, 99.5%, extra pure, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
500 mL
1 L
4 L
25 L
Packaging:
View all
Glass bottle
Catalog number 168312500
Price (USD)/ Each
30.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
35.80 
Save 5.15 (14%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
30.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
35.80 
Save 5.15 (14%)
Add to cart
Methacrylic acid, 99.5%, extra pure, stabilized, Thermo Scientific Chemicals
Catalog number168312500
Price (USD)/ Each
30.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
35.80 
Save 5.15 (14%)
-
Add to cart
Chemical Identifiers
CAS79-41-4
IUPAC Name2-methylprop-2-enoic acid
Molecular FormulaC4H6O2
InChI KeyCERQOIWHTDAKMF-UHFFFAOYSA-N
SMILESCC(=C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.4 %
Color scale=<20 APHA
Appearance (Form)Clear liquid
Refractive index1.4295 to 1.4315 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only