1x1 image pixel for data collection
2-Methyltetrahydrofuran, 99+%, pure, stabilized
2-Methyltetrahydrofuran, 99+%, pure, stabilized
2-Methyltetrahydrofuran, 99+%, pure, stabilized
2-Methyltetrahydrofuran, 99+%, pure, stabilized
Thermo Scientific Chemicals

2-Methyltetrahydrofuran, 99+%, pure, stabilized

CAS: 96-47-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 168365000
Price (USD)
179.65
Online Exclusive
199.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
179.65
Online Exclusive
199.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS96-47-9
IUPAC Name2-methyloxolane
Molecular FormulaC5H10O
InChI KeyJWUJQDFVADABEY-UHFFFAOYNA-N
SMILESCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Water=<0.2 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link