Tetrabutylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals
Tetrabutylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrabutylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 168380250
Price (USD)/ Each
48.65
Online exclusive
57.40 
Save 8.75 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
48.65
Online exclusive
57.40 
Save 8.75 (15%)
Add to cart
Tetrabutylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals
Catalog number168380250
Price (USD)/ Each
48.65
Online exclusive
57.40 
Save 8.75 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametetrabutylazanium hydrogen sulfate
Molecular FormulaC16H37NO4S
InChI KeySHFJWMWCIHQNCP-UHFFFAOYSA-M
SMILESOS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol)339.54
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point168°C to 173°C
Titration with NaOH>=97.5 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only