2-Fluoro-5-nitroaniline, 98%, Thermo Scientific Chemicals
2-Fluoro-5-nitroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoro-5-nitroaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 168390050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-fluoro-5-nitroaniline
Molecular FormulaC6H5FN2O2
InChI KeyKJVBJICWGQIMOZ-UHFFFAOYSA-N
SMILESNC1=CC(=CC=C1F)[N+]([O-])=O
Molecular Weight (g/mol)156.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Ochre to light brown
Melting point99°C to 103°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only