Thermo Scientific Chemicals

2-Fluoro-5-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 168391000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Fluoro-5-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 168391000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 168391000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
369-36-8
IUPAC Name
2-fluoro-5-nitroaniline
Molecular Formula
C6H5FN2O2
InChI Key
KJVBJICWGQIMOZ-UHFFFAOYSA-N
SMILES
NC1=CC(=CC=C1F)[N+]([O-])=O
Appearance (Color)
Ochre to light brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
99°C to 103°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text